Assay Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102458.1 | NP_001095928.1 | 10 - 11 | 1649 | 155 | |
| NM_001301688.1 | NP_001288617.1 | 9 - 10 | 1489 | 155 | ||
| NM_018745.5 | NP_061215.1 | 9 - 10 | 1489 | 155 | ||
| GenBank mRNA | AF032128.1 | - | 7 - 8 | 915 | 155 | |
| AK049680.1 | - | 9 - 10 | 1490 | 155 | ||
| AK088112.1 | - | 9 - 10 | 1487 | 155 | ||
| AK088671.1 | - | 10 - 11 | 1646 | 155 | ||
| AK133914.1 | - | 9 - 10 | 1485 | 155 | ||
| BC019412.1 | - | 9 - 10 | 1477 | 5151406 | 155 | |
| BC021152.1 | - | 9 - 10 | 1442 | 155 | ||
| BC043722.1 | - | 7 - 8 | 995 | 5321733 | 155 | |
| CF738062.1 | - | 5 - 6 | 480 | 155 | ||
More Information
Set Membership: |
|

Azin1
Genomic Map