Assay Details
| Gene Symbol: Klhl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161800.1 | NP_001155272.1 | 9 - 10 | 1547 | 105 | |
| GenBank mRNA | BC029154.1 | - | 9 - 10 | 1515 | 4952319 | 105 |
More Information
Set Membership: |
|

Klhl7
Genomic Map