Assay Details
| Gene Symbol: Vmn1r195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134223.2 | NP_598984.2 | 1 | 890 | 122 | |
| GenBank mRNA | BC116946.1 | - | 1 | 890 | 40126265 | 122 |
| BC119333.1 | - | 1 | 890 | 8734082 | 122 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154585 | NM_134223.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154585 | BC116946.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154585 | BC119333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154585 | NM_134223.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154585 | BC116946.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154585 | BC119333.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151083 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154585 | NM_134223.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154585 | BC116946.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154585 | BC119333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154585 | NM_134223.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154585 | BC116946.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154585 | BC119333.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154585 | NM_134223.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154585 | BC116946.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154585 | BC119333.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154585 | NM_134223.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154585 | BC116946.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154585 | BC119333.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28877066 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151083 | - | - |
| Whole Mouse Genome G4122A | A_51_P154585 | NM_134223.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154585 | BC116946.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154585 | BC119333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154585 | NM_134223.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154585 | BC116946.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154585 | BC119333.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131638 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151083 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969317 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969305 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969307 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28877021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10659303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10969305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10620806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10630593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10659315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10444370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28877076 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10403987 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403986 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300001271.mm.1 | NM_134223.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001271.mm.1 | NM_134223.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001271.mm.1 | TC1300000198.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001272.mm.1 | NM_134223.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001272.mm.1 | NM_134223.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001272.mm.1 | TC1300000198.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1740020 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740020 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237381 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237381 | - | - |
| mouse-6 v1.1 (Illumina) | scl12352.1.1_295-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740020 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl12352.1.1_295-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740020 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237381 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237381 | - | - |

Vmn1r195
Genomic Map