Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013646.1 | - | 1 - 2 | 60 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013646.1 transcripts.
Set Membership: |
|

Tmem219
Genomic Map