| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1432226_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1432226_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1432226_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1500002562.mm.1 |
NR_045811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002562.mm.1 |
NR_045811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002562.mm.1 |
TC1500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500002567.mm.1 |
NR_045811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002567.mm.1 |
NR_045811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002567.mm.1 |
TC1500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500002563.mm.1 |
NR_045811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002563.mm.1 |
NR_045811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002563.mm.1 |
TC1500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500002564.mm.1 |
NR_045811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002564.mm.1 |
NR_045811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002564.mm.1 |
TC1500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500002566.mm.1 |
NR_045811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002566.mm.1 |
NR_045811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002566.mm.1 |
TC1500000363.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1432226_at |
- |
- |
| Mouse Expression Set 430 |
1432226_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1432226_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1432226_PM_at |
- |
- |