Assay Details
| Gene Symbol: Fastk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023229.2 | NP_075718.2 | 9 - 10 | 1625 | 65 | |
| GenBank mRNA | AF168682.1 | - | 8 - 9 | 1520 | 65 | |
| AK002552.1 | - | 9 - 10 | 1573 | 65 | ||
| AK003940.1 | - | 6 - 7 | 868 | 65 | ||
| AK038596.1 | - | 9 - 10 | 1577 | 65 | ||
| AK052667.1 | - | 9 - 10 | 1570 | 65 | ||
| AK139374.1 | - | 8 - 9 | 2345 | 65 | ||
| AK151859.1 | - | 9 - 10 | 1577 | 65 | ||
| AK152369.1 | - | 9 - 10 | 1626 | 65 | ||
| AK159751.1 | - | 9 - 10 | 1577 | 65 | ||
| AK167346.1 | - | 8 - 9 | 2337 | 65 | ||
| BC013547.1 | - | 9 - 10 | 1542 | 4218374 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541891 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059337 | BC013547.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059337 | AK167346.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059337 | AK152369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059337 | AK159751.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059337 | AK139374.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145376 | AK166508.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145376 | AK164317.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344152 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164656 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059337 | BC013547.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059337 | AK167346.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059337 | AK152369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059337 | AK159751.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059337 | AK139374.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145376 | AK166508.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145376 | AK164317.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348764 | AF168682.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541887 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164656 | - | - |
| Whole Mouse Genome G4122A | A_51_P348764 | AF168682.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130973 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164656 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04131815 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04161721 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04131826 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04131818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04131808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04131822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04131815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04161721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04272652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04131809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04272658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541868 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10528608 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528607 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528605 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528609 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528606 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528602 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460635_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528598 | - | - |
| Mouse Genome 430 2.0 Array | 1460635_at | - | - |
| Mouse Genome 430 2.0 Array | 1460635_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66587_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66587_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500018897.mm.1 | NM_023229.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018897.mm.1 | NM_023229.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018897.mm.1 | TC0500002078.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460635_PM_at | - | - |
| Mouse Expression Set 430 | 1460635_at | - | - |
| Murine Genome U74v2 Set | 97800_PM_at | - | - |
| Murine Genome U74v2 Set | 97800_at | - | - |
| Murine Genome U74v2 Set | 167638_PM_r_at | - | - |
| Murine Genome U74v2 Set | 167638_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460635_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460635_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460635_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460635_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2690053 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4280288 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6370292 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4610551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4610551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2690053 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4280288 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370292 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002690053 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370292 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2644697 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2672662 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243206 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245926 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2672662 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245926 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243206 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2644697 | - | - |
| mouse-6 v1.1 (Illumina) | scl0004210.1_43-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003979.1_54-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0004207.1_37-S | - | - |
| mouse-6 v1.1 (Illumina) | scl26802.3_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2690053 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610551 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370292 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280288 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26802.3_29-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004210.1_43-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004207.1_37-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003979.1_54-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610551 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280288 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2690053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2847136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2672662 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2672662 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2847136 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0004207.1_37-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0004210.1_43-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl26802.3_29-S | - | - |

Fastk
Genomic Map