Assay Details
| Gene Symbol: Has1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008215.2 | NP_032241.1 | 1 - 2 | 53 | 58 | |
| GenBank mRNA | AK053726.1 | - | 1 - 2 | 71 | 58 | |
| BC125424.1 | - | 1 - 2 | 32 | 40129939 | 58 | |
| BC125426.1 | - | 1 - 2 | 32 | 40129941 | 58 | |
| D82964.1 | - | 1 - 2 | 59 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK053726.1, BC125424.1, BC125426.1, D82964.1, NM_008215.2 transcripts.
Set Membership: |
|

Has1
Genomic Map