Assay Details
| Gene Symbol: Mrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013389.2 | NP_001013407.2 | 4 - 5 | 459 | 132 | |
| GenBank mRNA | AW456688.1 | - | 4 - 5 | 285 | 132 | |
| CA752560.1 | - | 1 - 2 | 112 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrs2
Genomic Map