Assay Details
| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013791.2 | NP_038819.1 | 3 - 4 | 342 | 164 | |
| GenBank mRNA | AK037967.1 | - | 3 - 4 | 343 | 164 | |
| AK077541.1 | - | 3 - 4 | 338 | 164 | ||
| AK155457.1 | - | 3 - 4 | 323 | 164 | ||
| AK164822.1 | - | 3 - 4 | 338 | 164 | ||
| AK165239.1 | - | 3 - 4 | 342 | 164 | ||
| AK171980.1 | - | 3 - 4 | 326 | 164 | ||
| BC013703.1 | - | 3 - 4 | 326 | 3668925 | 164 | |
| U72194.1 | - | 3 - 4 | 325 | 164 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkln1
Genomic Map