Assay Details
| Gene Symbol: Tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025982.4 | NP_080258.1 | 4 - 5 | 526 | 82 | |
| GenBank mRNA | AK012457.1 | - | 4 - 5 | 527 | 82 | |
| AK012567.1 | - | 4 - 5 | 529 | 82 | ||
| AK012853.1 | - | 4 - 5 | 529 | 82 | ||
| AK014183.1 | - | 4 - 5 | 525 | 82 | ||
| AK075962.1 | - | 4 - 5 | 527 | 82 | ||
| AK160269.1 | - | 2 - 3 | 1136 | 82 | ||
| BC023306.1 | - | 4 - 5 | 503 | 4165774 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan31
Genomic Map