Assay Details
| Gene Symbol: Fbxo43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081253.1 | NP_001074722.1 | 4 - 5 | 2202 | 75 | |
| GenBank mRNA | AK019714.1 | - | 4 - 5 | 2140 | 75 | |
| AK030012.1 | - | 4 - 5 | 2023 | 75 | ||
| BC026503.1 | - | 3 - 4 | 496 | 75 | ||
| BC098484.1 | - | 4 - 5 | 2202 | 30053208 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975655 | BC026503.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975655 | NM_001081253.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768820 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809989 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135628 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126812 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156058 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166786 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975655 | BC026503.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975655 | NM_001081253.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126812 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156058 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166786 | - | - |
| Whole Mouse Genome G4122A | A_52_P591353 | BC026503.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P591353 | NM_001081253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P260613 | BC098484.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P260613 | AK030012.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P260613 | BC026503.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P260613 | NM_001081253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P260613 | AK019714.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126812 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156058 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166786 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12361112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12080302 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12349098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12001314 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12349102 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12080304 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12001311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12095944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12337113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12320088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11978112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12188112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11978111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12112176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12320087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12464995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12396923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12275923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12349903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12112177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12349102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12349105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30136916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11897903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12203422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11985956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137031 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10428155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428156 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428152 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427415_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10428150 | - | - |
| Mouse Genome 430 2.0 Array | 1427415_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427415_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78803_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78803_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500010553.mm.1 | NM_001081253.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010553.mm.1 | NM_001081253.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010553.mm.1 | TC1500001368.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500010552.mm.1 | NM_001081253.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010552.mm.1 | NM_001081253.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010552.mm.1 | TC1500001368.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427415_at | - | - |
| Mouse Expression Set 430 | 1427415_PM_at | - | - |
| Murine Genome U74v2 Set | 166668_PM_at | - | - |
| Murine Genome U74v2 Set | 166668_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427415_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427415_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427415_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427415_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106020026 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734262 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2432774 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734262 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2734262 | - | - |

Fbxo43
Genomic Map