Assay Details
| Gene Symbol: Olfml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172907.3 | NP_766495.1 | 1 - 2 | 457 | 79 | |
| GenBank mRNA | AK032948.1 | - | 1 - 2 | 418 | 79 | |
| AK140277.1 | - | 1 - 2 | 210 | 79 | ||
| BC047207.1 | - | 1 - 2 | 444 | 6310165 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Olfml1
Genomic Map