Assay Details
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015083.1 | - | 5 - 6 | 619 | 122 | |
AK157931.1 | - | 429 | 122 | |||
BY752674.1 | - | 4 - 5 | 429 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
