Assay Details
| Gene Symbol: Usp31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033173.1 | NP_001028345.1 | 11 - 12 | 1808 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp31| Gene Symbol: Usp31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033173.1 | NP_001028345.1 | 11 - 12 | 1808 | 81 | |
Set Membership: |
|