Assay Details
| Gene Symbol: Map7d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081124.1 | NP_001074593.1 | 12 - 13 | 1874 | 76 | |
| GenBank mRNA | AK030316.1 | - | 8 - 9 | 1376 | 76 | |
| AK045412.1 | - | 5 - 6 | 850 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map7d2
Genomic Map