Assay Details
| Gene Symbol: Map7d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081124.1 | NP_001074593.1 | 2 - 3 | 396 | 81 | |
| GenBank mRNA | AK013466.1 | - | 2 - 3 | 187 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map7d2
Genomic Map