Assay Details
| Gene Symbol: Map7d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081124.1 | NP_001074593.1 | 8 - 9 | 1127 | 85 | |
| GenBank mRNA | AK013466.1 | - | 8 - 9 | 916 | 85 | |
| AK019929.1 | - | 4 - 5 | 725 | 85 | ||
| AK030316.1 | - | 4 - 5 | 629 | 85 | ||
| AK045412.1 | - | 1 - 2 | 103 | 85 | ||
| AK162227.1 | - | 5 - 6 | 562 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map7d2
Genomic Map