Assay Details
| Gene Symbol: Map7d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081124.1 | NP_001074593.1 | 10 - 11 | 1552 | 76 | |
| GenBank mRNA | AK005554.1 | - | 6 - 7 | 1068 | 76 | |
| AK030316.1 | - | 6 - 7 | 1054 | 76 | ||
| AK043342.1 | - | 6 - 7 | 1127 | 76 | ||
| AK045412.1 | - | 3 - 4 | 528 | 76 | ||
| AK162227.1 | - | 7 - 8 | 987 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map7d2
Genomic Map