Assay Details
| Gene Symbol: Calu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285412.1 | NP_001272341.1 | 3 - 4 | 343 | 152 | |
| NM_007594.4 | NP_031620.1 | 3 - 4 | 526 | 152 | ||
| GenBank mRNA | AK135295.1 | - | 3 - 4 | 451 | 152 | |
| AK148178.1 | - | 3 - 4 | 479 | 152 | ||
| AK160795.1 | - | 3 - 4 | 479 | 152 | ||
| AK161807.1 | - | 3 - 4 | 483 | 152 | ||
| AK163674.1 | - | 3 - 4 | 514 | 152 | ||
| BC051423.1 | - | 3 - 4 | 487 | 3989906 | 152 | |
| U81829.1 | - | 3 - 4 | 452 | 152 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285412.1 transcripts.
Set Membership: |
|

Calu
Genomic Map