Assay Details
| Gene Symbol: Spesp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025721.2 | NP_079997.1 | 1 - 2 | 175 | 60 | |
| GenBank mRNA | AK014843.1 | - | 1 - 2 | 176 | 60 | |
| AK015620.1 | - | 1 - 2 | 206 | 60 | ||
| BC050754.1 | - | 1 - 2 | 183 | 6773961 | 60 | |
More Information
Set Membership: |
|

Spesp1
Genomic Map