Assay Details
| Gene Symbol: Sik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027498.3 | NP_081774.3 | 6 - 7 | 900 | 71 | |
| GenBank mRNA | AK017789.1 | - | 5 - 6 | 650 | 71 | |
| BC063268.1 | - | 6 - 7 | 696 | 4187769 | 71 | |
| BC080688.1 | - | 6 - 7 | 842 | 6515742 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sik3
Genomic Map