Assay Details
| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030256.2 | NP_084532.2 | 1 - 2 | 133 | 110 | |
| GenBank mRNA | AY296058.1 | - | 1 - 2 | 133 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY296058.1, NM_030256.2 transcripts.
Set Membership: |
|

Bcl9l
Genomic Map