Assay Details
| Gene Symbol: Prtn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011178.2 | NP_035308.2 | 2 - 3 | 244 | 72 | |
| GenBank mRNA | U43525.1 | - | 2 - 3 | 232 | 72 | |
| U97073.1 | - | 2 - 3 | 244 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prtn3
Genomic Map