Assay Details
| Gene Symbol: Nlrp9c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042612.1 | NP_001036077.1 | 3 - 4 | 2024 | 139 | |
| GenBank mRNA | AK145425.1 | - | 2 - 3 | 1637 | 139 | |
| AK163321.1 | - | 3 - 4 | 1794 | 139 | ||
| AY596203.1 | - | 3 - 4 | 2024 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107578 | AY360472.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056714 | AY596203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056714 | AK145425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056714 | NM_001042612.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424978 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105283 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 797235 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23012 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146037 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164265 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145199 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056714 | AY596203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056714 | AK145425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056714 | NM_001042612.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107578 | AY360472.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06145354 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164265 | - | - |
| Whole Mouse Genome G4122A | A_51_P276203 | AK087873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P167935 | AY596203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P167935 | AY360472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P167935 | AK145425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P167935 | NM_001042612.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164265 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06411952 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06411951 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06164527 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06411950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25013779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06411953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06311204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06130103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06403487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06468531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06130108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25013802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06363641 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561145 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439636_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439217_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561142 | - | - |
| Mouse Genome 430 2.0 Array | 1439217_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439636_at | - | - |
| Mouse Genome 430 2.0 Array | 1439217_at | - | - |
| Mouse Genome 430 2.0 Array | 1439636_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330490_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330490_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022961.mm.1 | NM_001042612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022961.mm.1 | NM_001042612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022961.mm.1 | TC0700002650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022956.mm.1 | NM_001042612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022956.mm.1 | NM_001042612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022956.mm.1 | TC0700002650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022962.mm.1 | NM_001042612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022962.mm.1 | NM_001042612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022962.mm.1 | TC0700002650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022959.mm.1 | NM_001042612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022959.mm.1 | NM_001042612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022959.mm.1 | TC0700002650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022957.mm.1 | NM_001042612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022957.mm.1 | NM_001042612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022957.mm.1 | TC0700002650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022958.mm.1 | NM_001042612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022958.mm.1 | NM_001042612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022958.mm.1 | TC0700002650.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439217_at | - | - |
| Mouse Expression Set 430 | 1439636_at | - | - |
| Mouse Expression Set 430 | 1439217_PM_at | - | - |
| Mouse Expression Set 430 | 1439636_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439217_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439217_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439636_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439636_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 792463 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2646906 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2646906 | - | - |

Nlrp9c
Genomic Map