Assay Details
| Gene Symbol: Adamts14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081127.1 | NP_001074596.1 | 14 - 15 | 2343 | 60 | |
| GenBank mRNA | BC157953.1 | - | 14 - 15 | 2178 | 9007283 | 60 |
| BC158014.1 | - | 14 - 15 | 2176 | 9088151 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamts14
Genomic Map