Assay Details
| Gene Symbol: Adamts14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081127.1 | NP_001074596.1 | 3 - 4 | 843 | 61 | |
| GenBank mRNA | AK145103.1 | - | 3 - 4 | 845 | 61 | |
| BC157953.1 | - | 3 - 4 | 678 | 9007283 | 61 | |
| BC158014.1 | - | 3 - 4 | 676 | 9088151 | 61 | |
| BC172106.1 | - | 3 - 4 | 678 | 61 | ||
More Information
Set Membership: |
|

Adamts14
Genomic Map