Assay Details
| Gene Symbol: Pbld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026701.2 | NP_080977.2 | 181 | 61 | ||
| GenBank mRNA | AK002802.1 | - | 2 - 3 | 201 | 61 | |
| AK143947.1 | - | 2 - 3 | 217 | 61 | ||
| BC019997.1 | - | 181 | 4240949 | 61 | ||
More Information
Set Membership: |
|

Pbld1
Genomic Map