Assay Details
| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025615.3 | NP_001020786.1 | 1 - 2 | 177 | 67 | |
| NM_001289436.1 | NP_001276365.1 | 1 - 2 | 177 | 67 | ||
| NM_026202.4 | NP_080478.2 | 1 - 2 | 177 | 67 | ||
| GenBank mRNA | AJ534985.1 | - | 1 - 2 | 44 | 67 | |
| AK012190.1 | - | 1 - 2 | 71 | 67 | ||
| AK016827.1 | - | 1 - 2 | 152 | 67 | ||
| AK077557.1 | - | 1 - 2 | 115 | 67 | ||
| AK147820.1 | - | 1 - 2 | 128 | 67 | ||
| AK151672.1 | - | 1 - 2 | 139 | 67 | ||
| AK162434.1 | - | 1 - 2 | 154 | 67 | ||
| AK165214.1 | - | 1 - 2 | 82 | 67 | ||
| BC144895.1 | - | 1 - 2 | 156 | 9053443 | 67 | |
| BC144898.1 | - | 1 - 2 | 156 | 67 | ||
| BC145933.1 | - | 1 - 2 | 156 | 40131236 | 67 | |
| BC145935.1 | - | 1 - 2 | 156 | 40131238 | 67 | |
| BY716501.1 | - | 1 - 2 | 152 | 67 | ||
| CJ045424.1 | - | 1 - 2 | 181 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001289436.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc50
Genomic Map