Assay Details
| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289436.1 | NP_001276365.1 | 7 - 8 | 742 | 93 | |
| NM_026202.4 | NP_080478.2 | 6 - 7 | 696 | 93 | ||
| GenBank mRNA | AJ534985.1 | - | 6 - 7 | 563 | 93 | |
| AK012190.1 | - | 6 - 7 | 590 | 93 | ||
| AK147820.1 | - | 6 - 7 | 93 | |||
| AK151672.1 | - | 6 - 7 | 658 | 93 | ||
| AK162434.1 | - | 6 - 7 | 673 | 93 | ||
| BC144898.1 | - | 7 - 8 | 722 | 93 | ||
| BC145933.1 | - | 6 - 7 | 675 | 40131236 | 93 | |
| BC145935.1 | - | 6 - 7 | 675 | 40131238 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc50
Genomic Map