| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600003619.mm.1 |
NM_001025615.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003619.mm.1 |
NM_001289436.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003619.mm.1 |
NM_026202.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003619.mm.1 |
TC1600000379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600003620.mm.1 |
NM_001025615.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003620.mm.1 |
NM_001289436.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003620.mm.1 |
NM_026202.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003620.mm.1 |
TC1600000379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600003622.mm.1 |
NM_001025615.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003622.mm.1 |
NM_001289436.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003622.mm.1 |
NM_026202.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600003622.mm.1 |
TC1600000379.mm.1 |
MTA TC |