Assay Details
| Gene Symbol: Umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009471.2 | NP_033497.1 | 2 - 3 | 336 | 73 | |
| GenBank mRNA | AK035821.1 | - | 2 - 3 | 337 | 73 | |
| AK090102.1 | - | 2 - 3 | 339 | 73 | ||
| BC003887.1 | - | 2 - 3 | 333 | 3590810 | 73 | |
| BC095981.1 | - | 2 - 3 | 322 | 3499037 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Umps
Genomic Map