Assay Details
| Gene Symbol: Ndufb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026610.1 | NP_080886.1 | 2 - 3 | 332 | 113 | |
| GenBank mRNA | AK009807.1 | - | 2 - 3 | 332 | 113 | |
| AK018707.1 | - | 2 - 3 | 332 | 113 | ||
| BC056229.1 | - | 2 - 3 | 333 | 6475281 | 113 | |
| BC096421.1 | - | 2 - 3 | 331 | 113 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30681785 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30681787 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30681786 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30681782 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965539 | AK009807.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965539 | AK018707.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965539 | BC056229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965539 | BC096421.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965539 | NM_026610.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P123527 | AK018707.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P123527 | NM_026610.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85779 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117452 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P123527 | AK018707.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P123527 | NM_026610.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965539 | AK009807.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965539 | AK018707.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965539 | BC056229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965539 | BC096421.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965539 | NM_026610.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434269 | AK009807.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434269 | AK018707.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434269 | BC056229.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434269 | BC087907.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434269 | BC096421.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30681790 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08567292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117452 | - | - |
| Whole Mouse Genome G4122A | A_51_P434269 | AK009807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434269 | AK018707.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434269 | BC056229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434269 | BC087907.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434269 | BC096421.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126298 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123527 | AK018707.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123527 | NM_026610.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117452 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12541247 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12541259 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12541252 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30681782 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30681786 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30681787 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30681785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30681760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12675017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12541263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12952214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12742534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12777565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12777564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13022977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12952213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12683269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30681731 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10439400 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10462194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439401 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432427_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10462193 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10439398 | - | - |
| Mouse Genome 430 2.0 Array | 1432427_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428075_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428075_at | - | - |
| Mouse Genome 430 2.0 Array | 1432427_at | - | - |
| Mouse Genome 430 2.0 Array | 1428076_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1428076_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600013051.mm.1 | NM_026610.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013051.mm.1 | NM_026610.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013051.mm.1 | TC1600001593.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428076_s_at | - | - |
| Mouse Expression Set 430 | 1428075_PM_at | - | - |
| Mouse Expression Set 430 | 1428076_PM_s_at | - | - |
| Mouse Expression Set 430 | 1428075_at | - | - |
| Murine Genome U74v2 Set | 94541_PM_at | - | - |
| Murine Genome U74v2 Set | 94541_at | - | - |
| Murine Genome U74v2 Set | 171510_PM_i_at | - | - |
| Murine Genome U74v2 Set | 171510_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428076_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428076_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428075_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428075_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432427_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432427_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 832775 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5690594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5690594 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227012 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227012 | - | - |
| mouse-6 v1.1 (Illumina) | scl0068194.2_330-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690594 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068194.2_330-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690594 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1227012 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227012 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0068194.2_330-S | - | - |

Ndufb4
Genomic Map