Assay Details
Gene Symbol: Rfc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145480.1 | NP_663455.1 | 6 - 7 | 610 | 119 | |
GenBank mRNA | AK146954.1 | - | 6 - 7 | 598 | 119 | |
AK147030.1 | - | 6 - 7 | 598 | 119 | ||
BC003335.1 | - | 6 - 7 | 610 | 3257208 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
