Assay Details
| Gene Symbol: Rfc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145480.1 | NP_663455.1 | 8 - 9 | 857 | 97 | |
| GenBank mRNA | AK146954.1 | - | 8 - 9 | 845 | 97 | |
| AK147030.1 | - | 8 - 9 | 845 | 97 | ||
| BC003335.1 | - | 8 - 9 | 857 | 3257208 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfc4
Genomic Map