| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600004229.mm.1 |
NM_080457.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004229.mm.1 |
NM_080457.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004229.mm.1 |
TC1600000450.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004233.mm.1 |
NM_080457.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004233.mm.1 |
NM_080457.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004233.mm.1 |
TC1600000450.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004235.mm.1 |
NM_080457.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004235.mm.1 |
NM_080457.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004235.mm.1 |
TC1600000450.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004231.mm.1 |
NM_080457.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004231.mm.1 |
NM_080457.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004231.mm.1 |
TC1600000450.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004234.mm.1 |
NM_080457.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004234.mm.1 |
NM_080457.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004234.mm.1 |
TC1600000450.mm.1 |
MTA TC |