Assay Details
| Gene Symbol: Zdhhc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172151.4 | NP_742163.4 | 10 - 11 | 2245 | 75 | |
| GenBank mRNA | AK032069.1 | - | 1 - 2 | 235 | 75 | |
| AY668947.1 | - | 10 - 11 | 2119 | 75 | ||
| AY894892.1 | - | 10 - 11 | 2245 | 75 | ||
| BC055694.1 | - | 1 - 2 | 542 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zdhhc8
Genomic Map