Assay Details
| Gene Symbol: Zdhhc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172151.4 | NP_742163.4 | 1 - 2 | 231 | 74 | |
| GenBank mRNA | AY668947.1 | - | 1 - 2 | 105 | 74 | |
| AY894892.1 | - | 1 - 2 | 231 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc8
Genomic Map