Assay Details
| Gene Symbol: Zdhhc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172151.4 | NP_742163.4 | 6 - 7 | 879 | 89 | |
| GenBank mRNA | AY668947.1 | - | 6 - 7 | 753 | 89 | |
| AY894892.1 | - | 6 - 7 | 879 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zdhhc8
Genomic Map