Assay Details
| Gene Symbol: Mpzl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093749.2 | NP_001087218.1 | 3 - 4 | 513 | 82 | |
| GenBank mRNA | EF102773.1 | - | 3 - 4 | 477 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpzl3
Genomic Map