Assay Details
| Gene Symbol: Pldi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033616.1 | 1 - 2 | 198 | 77 | ||
| GenBank mRNA | AK007277.1 | - | 1 - 2 | 201 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007277.1 transcripts.
Set Membership: |
|

Pldi
Genomic Map