Assay Details
| Gene Symbol: Apba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018758.2 | NP_061228.1 | 7 - 8 | 1439 | 75 | |
| GenBank mRNA | AF070975.1 | - | 7 - 8 | 1393 | 75 | |
| AK082046.1 | - | 1359 | 75 | |||
| AK169462.1 | - | 7 - 8 | 1423 | 75 | ||
| AK170182.1 | - | 7 - 8 | 1387 | 75 | ||
| BC005423.1 | - | 7 - 8 | 1371 | 3487448 | 75 | |
| BC009666.1 | - | 7 - 8 | 1383 | 3490710 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apba3
Genomic Map