Assay Details
| Gene Symbol: Lsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164184.1 | NP_001157656.1 | 3 - 4 | 859 | 85 | |
| GenBank mRNA | AK146807.1 | - | 3 - 4 | 575 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lsr
Genomic Map