Assay Details
| Gene Symbol: Gcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008103.3 | NP_032129.2 | 1 - 2 | 79 | 72 | |
| GenBank mRNA | AK142574.1 | - | 1 - 2 | 65 | 72 | |
| D88612.1 | - | 1 - 2 | 114 | 72 | ||
| U59876.1 | - | 1 - 2 | 95 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142574.1, D88612.1, NM_008103.3, U59876.1 transcripts.
Set Membership: |
|

Gcm1
Genomic Map