Assay Details
| Gene Symbol: Gldn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177350.5 | NP_796324.1 | 5 - 6 | 722 | 121 | |
| GenBank mRNA | AF548022.1 | - | 5 - 6 | 378 | 121 | |
| AK031523.1 | - | 5 - 6 | 723 | 121 | ||
| BC125025.1 | - | 5 - 6 | 702 | 40058551 | 121 | |
| BC125026.1 | - | 5 - 6 | 702 | 40058552 | 121 | |
More Information
Set Membership: |
|

Gldn
Genomic Map