Assay Details
| Gene Symbol: Ctnna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164376.1 | NP_001157848.1 | 13 - 14 | 2040 | 128 | |
| NM_001164519.1 | NP_001157991.1 | 8 - 9 | 1199 | 128 | ||
| NM_177612.3 | NP_808280.2 | 13 - 14 | 1968 | 128 | ||
| GenBank mRNA | AF344871.1 | - | 13 - 14 | 2040 | 128 | |
| AK030166.1 | - | 13 - 14 | 1969 | 128 | ||
| AK142745.1 | - | 13 - 14 | 2041 | 128 | ||
| BC112409.1 | - | 13 - 14 | 1942 | 40060582 | 128 | |
| DQ380429.1 | - | 13 - 14 | 1968 | 128 | ||
| DQ380430.1 | - | 8 - 9 | 1198 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnna3
Genomic Map