Assay Details
| Gene Symbol: Ctnna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164376.1 | NP_001157848.1 | 15 - 16 | 2314 | 77 | |
| NM_001164519.1 | NP_001157991.1 | 10 - 11 | 1473 | 77 | ||
| NM_177612.3 | NP_808280.2 | 15 - 16 | 2242 | 77 | ||
| GenBank mRNA | AF344871.1 | - | 15 - 16 | 2314 | 77 | |
| AK030166.1 | - | 15 - 16 | 2243 | 77 | ||
| AK142745.1 | - | 15 - 16 | 2315 | 77 | ||
| BC112409.1 | - | 15 - 16 | 2216 | 40060582 | 77 | |
| DQ380429.1 | - | 15 - 16 | 2242 | 77 | ||
| DQ380430.1 | - | 10 - 11 | 1472 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnna3
Genomic Map