Assay Details
| Gene Symbol: Lca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027448.2 | NP_081724.1 | 7 - 8 | 1600 | 73 | |
| GenBank mRNA | AK017509.1 | - | 2 - 3 | 174 | 73 | |
| BC049101.1 | - | 7 - 8 | 1528 | 5702365 | 73 | |
| BC052060.1 | - | 7 - 8 | 1528 | 5702365 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lca5
Genomic Map