Assay Details
| Gene Symbol: Nxpe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030069.3 | NP_084345.2 | 5 - 6 | 1435 | 94 | |
| GenBank mRNA | AK019469.1 | - | 5 - 6 | 1335 | 94 | |
| AK033667.1 | - | 5 - 6 | 1200 | 94 | ||
| AK157096.1 | - | 5 - 6 | 1373 | 94 | ||
| AK162300.1 | - | 5 - 6 | 1197 | 94 | ||
| AK165213.1 | - | 4 - 5 | 605 | 94 | ||
| BC140955.1 | - | 5 - 6 | 1318 | 9055567 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406165 | AK019469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406165 | AK019469.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121319 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104144 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406165 | AK019469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406165 | AK019469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406165 | AK019469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406165 | AK019469.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07833128 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07833138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07876374 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07833133 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07876373 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07833126 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07833129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26491343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07966532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07737107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07833126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07839332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07586027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07814269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08054765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07556738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08054768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08054742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07947502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07693734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07529428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07930550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07805060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08059297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07930544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07805059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07620990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07464832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08054764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07737113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26491341 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10593202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593200 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453220_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444471_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10593198 | - | - |
| Mouse Genome 430 2.0 Array | 1444471_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444471_at | - | - |
| Mouse Genome 430 2.0 Array | 1453220_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453220_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78252_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78252_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900018620.mm.1 | NM_030069.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018620.mm.1 | NM_030069.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018620.mm.1 | TC0900002233.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900018619.mm.1 | NM_030069.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018619.mm.1 | NM_030069.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018619.mm.1 | TC0900002233.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900018621.mm.1 | NM_030069.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018621.mm.1 | NM_030069.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018621.mm.1 | TC0900002233.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1444471_at | - | - |
| Mouse Expression Set 430 | 1453220_PM_at | - | - |
| Mouse Expression Set 430 | 1444471_PM_at | - | - |
| Mouse Expression Set 430 | 1453220_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444471_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444471_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453220_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453220_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 651503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3060088 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003060088 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770236 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2770236 | - | - |
| mouse-6 v1.1 (Illumina) | scl35908.8_513-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3060088 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35908.8_513-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060088 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2931918 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770236 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2931918 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl35908.8_513-S | - | - |

Nxpe2
Genomic Map