Assay Details
| Gene Symbol: Ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286546.1 | NP_001273475.1 | 5 - 6 | 633 | 62 | |
| GenBank mRNA | AK032645.1 | - | 5 - 6 | 519 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccpg1
Genomic Map