Assay Details
| Gene Symbol: Ubap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111145.1 | NP_001104615.1 | 2 - 3 | 259 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubap1l| Gene Symbol: Ubap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111145.1 | NP_001104615.1 | 2 - 3 | 259 | 92 | |
Set Membership: |
|